Switch & Go 8 Hour Multi Plug PAPR, Jetstream®, JSP LTD
Lieferant: JSP
CBB610-211-100
JSPACBB610-211-100EA 1770 EUR
JSPACBB610-211-100
Switch & Go 8 Hour Multi Plug PAPR, Jetstream®, JSP LTD
Respirators Atemschutz-Systeme
Jetstream® is a belt-mounted rechargeable respirator with a large capacity disposable dust filter. Jetstream® delivers 180 L/min. of air through the filter via a highly flexible hose to the desired headtop.
  • Belt-mounted rechargeable respirator
  • Large capacity disposable dust filter
  • No face fit required
  • PSL filter
The Jetstream® waist belt and power unit weighs just 765g. The Switch & Go unit comes complete with a white nylon head top. The system offers an assigned protection factor (APF) of 20 x Workplace Exposure Limit (WEL) for very fine dusts, fibres, fumes, and aqueous mists.
Standards: EN 12941:1998.
Lieferumfang: Jetstream® Switch & Go pack contains: PSL filter, hose, charger, carry case, battery, white nylon headtop, flow meter, and multi-region plug.