HEATING BATH 1500, USABLE FILLING 4 WITH DIGITAL TEMPERATURE CONTROL + 20 + 260 C, HEATING CAPACITY 230V/2000W DIMENSIONS CABINET: 250 X 250 X 60 MM POT DIAMETER. / H. 240 MM / 150 MM DATE 23/2015 1 * 1 ST
Lieferant: JUCHHEIM SOLINGEN
HEATING BATH 1500, USABLE FILLING 4 WITH DIGITAL TEMPERATURE CONTROL + 20 + 260 C, HEATING CAPACITY 230V/2000W DIMENSIONS CABINET: 250 X 250 X 60 MM POT DIAMETER. / H. 240 MM / 150 MM DATE 23/2015 1 * 1 ST