Double bossheads, cross shape
Lieferant: JUCHHEIM Laborgeräte GmbH
VL04850 VL04850S
JUCHVL04850EA 20.94 EUR
JUCHVL04850 JUCHVL04850S
Double bossheads, cross shape
Klemmen Stativmuffen
Bossheads made from either epoxy coated malleable iron or from stainless steel.