Biohazard Waste Cans
Lieferant: JUSTRITE MANUFACTURING
05930R
JUSU05930REA 134 EUR
JUSU05930R
Biohazard Waste Cans
Behälter Abfallbehälter
Leakproof waste cans with self-closing lids provide safe, temporary collection of biohazardous waste.
  • Great for medical facilities and research labs
  • Foot-operated, self-closing lid reduces risk of contamination
  • Reduce closure noise with SoundGard™ models
  • Sturdy design with steel construction
Safely contain biohazardous materials, including bloodborne pathogens and other potentially infectious materials (no sharps).
Meets OSHA 29 CFR 1910.1030 for the segregation of infectious waste and FM approved for use with alcohols and other flammable liquids.
Standards: FM, TUV, OSHA.
Bestellinformation: Includes OSHA biohazard label on front; use optional label kit 25880 for additional labels to add to the sides, to meet California Health and Safety Code.