Poly Safety Cans, Type I
Lieferant: JUSTRITE MANUFACTURING
12162
JUSU12162EA 137 EUR
JUSU12162
Poly Safety Cans, Type I
Sicherheitsbehälter
Chemical-resistance HDPE cans offer safe storage for flammable corrosives.
Standards: NFPA, OSHA.