Nonmetallic shelf cans have a faucet that is mounted two inches above the bottom to eliminate lifting the cans during dispensing
Nonmetallic tilt cans have faucet on top and the can rests in a sturdy metal cradle
Current-carrying carbon insert imbedded into the container provides proper grounding
Stainless steel hardware on the top of can provides maximum corrosion resistance and long life
These nonmetallic safety cans are ideal for dispensing the most corrosive liquids. It has a self-closing faucet for easy dispensing and all fill spouts have a pressure relief cap with stainless steel flame arrester.