Skywalker® 453 II PUR Anti-static Mat ESD
Lieferant: JUSTRITE MANUFACTURING
453-KNIEMATTE
JUSU453-KNIEMATTEEA 38.8 EUR
JUSU453-KNIEMATTE
Skywalker® 453 II PUR Anti-static Mat ESD
Fußmatten Permanente Bodenmatten
High-quality polyurethane foam anti-fatigue mats are reputed for their ultimate comfort in stationary standing positions. Its key characteristic of high thermal insulation performance is owed to a uniform closed cell structure wherein gas remains trapped.
  • Medium duty
  • R11 anti-slip pebble pattern
  • ESD static dissipative
  • Available in six standard sizes
  • Made from polyurethane foam
The integral skin adds to the compression force deflection properties (>120 according to ASTM 3574C), provides resistance to wear and is non-porous. Ideal for stationary standing workstations, sit-stand desks, packing stations, office environments, reception areas, wait stations, service counters, cashiers.