Aluminized Neck Protection
Lieferant: JUTEC
HSN030KA-1
JUTEHSN030KA-1EA 57.4 EUR
JUTEHSN030KA-1
Aluminized Neck Protection
Arbeitskleidung
Neck protection made of preox-aramide fabric.
  • Fabric resistant to flying sparks
  • Flexible, lightweight and soft
  • Including Velcro fastener
  • Suitable for Schuberth and Voss helmets
  • Universal dimensions
  • Weight: ~260 g/m²
Standards: EN ISO 11612:2015.