Plasmid Midiprep Kit, GeneJET™

Lieferant: Thermo Fisher Scientific

K0482 K0481
K0482EA 756 EUR
K0482 K0481
Plasmid Midiprep Kit, GeneJET™
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure

GeneJET™ Plasmid Midiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. The kit utilises silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 200 µg of high copy number plasmid DNA. The purified plasmid DNA can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.


  • Efficient: High yields up to 200 µg of high quality plasmid DNA from 50 ml of bacterial culture
  • High concentration of DNA purified: >400 ng/µl
  • Convenient: Spin column format, centrifuge or vacuum protocols
  • Fast: Procedure takes 60 minutes, considerably faster than using gravity flow columns


Lieferumfang: Each kit includes resuspension solution, lysis solution, neutralisation solution, endotoxin binding reagent, Wash Solution I (concentrated), Wash Solution II (concentrated), RNase A solution, elution buffer (10 mM Tris-HCl, pH 8,5), GeneJET™ Midi purification columns pre-assembled with collection tubes (15 ml), and collection tubes (15 ml).

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR