VWR®, DNA marker, 1 kb
Lieferant: VWR Chemicals
K181-1MLEA 275 EUR
K181-1ML
VWR®, DNA marker, 1 kb
Elektrophorese-Reagenzien Molekulargewichtsmarker
DNA marker consists of 11 fragments ranging from 500 to 10000 bp for easy band identification.
  • Sufficient for 150 lanes
  • Available pre-mixed with loading dye or with EZ-Vision® One DNA dye