Methylcellulose, 4000 mPa.S (2% H₂O), hochrein
Lieferant: VWR Chemicals
Synonyme:
Cellulose methyl ether
K390-250GEA
750
EUR
K390-250G
Methylcellulose, 4000 mPa.S (2% H₂O), hochrein
Methyl cellulose
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00081763 CAS-Nummer: 9004-67-5 |
Spezifikation Testergebnisse
Solubility (2%, Water) | Pass |
Viscosität (2%, Wasser)@20°C | 3500 - 5600 cps |
Appearance | white powder with a yellow cast |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...