Lithiumchlorid 8 mol/l in wässriger Lösung für die Biotechnologie

Lieferant: VWR Chemicals
Achtung

K445-100MLEA 249 EUR
K445-100ML
Lithiumchlorid 8 mol/l in wässriger Lösung für die Biotechnologie
Lithium chloride

Eine geeignete Fertiglösung, die Inhibitoren der cDNA Synthese und Translation beseitigt.


  • High concentration for easy dilution
  • Nuclease and protease free
  • Does not precipitate DNA, protein, or carbohydrates


This Lithium Chloride Solution comes especially formulated for RT PCR preparation and cDNA synthesis. This solution is a nuclease-free, protease-free, 8 Molar preparation of lithium chloride in deionised water and is highly concentrated for easy dilution.


It is ideal for the precipitation of RNA right after total RNA isolation or in vitro transcription. The lithium chloride is the key to this process, as it efficiently precipitates the RNA, but does not precipitate the DNA, protein, or free nucleotide triphosphates.


To ensure a quality product throughout, this 8 M Lithium Chloride Solution is 0.2 micron filtered and packaged in a PETG Nalge bottle. Solution is nuclease and protease free, and it is ready-to-use.

Formel: LiCl
Molecular Weight: 42,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011078
CAS-Nummer: 7447-41-8
EINECS: 231-212-3

Bestellen

Spezifikation Testergebnisse

DNasen Keine nachgewiesen
Proteasen Keine nachgewiesen
RNasen Keine nachgewiesen
Titration 7,7 - 8,2 Molar
Aussehen Klare, farblose Flüssigkeit

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR