Ammoniumperoxodisulfat (APS) 150 mg, Tabletten, Biotechnologische Qualität

Lieferant: VWR Chemicals

K833-100TABSEA 152 EUR
K833-100TABS
Ammoniumperoxodisulfat (APS) 150 mg, Tabletten, Biotechnologische Qualität
Ammoniumperoxodisulfat (APS)

Ammonium persulphate forms free radicals when dissolved in water, which initiate polymerisation of acrylamide solutions.


  • Convenient tablet, easily prepared without weighing
  • Fast and easy polymerisation of acrylamide mixtures
  • Odour-free and non flammable
  • Non hazardous shipping


Due to the instability of APS in water, a working solution must be prepared daily for optimum performance. A 10% APS solution can be easily be prepared, without weighing, by adding one 150 mg APS tablet to 1,5 ml of water.

Formel: (NH₄)₂S₂O₈
Lagertemperatur: Raumtemperatur
MDL: MFCD00003390
CAS-Nummer: 7727-54-0

Bestellen

Spezifikation Testergebnisse

Zerfallszeit 4 Minuten
Löslichkeit (1 Tablette/1 ml Wasser)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR