PBS/Tween® Pufferkonzentrat, 20X

Lieferant: VWR Chemicals

K875-500MLEA 308 EUR
K875-500ML
PBS/Tween® Pufferkonzentrat, 20X
Puffer Elektrophorese-Puffer Waschpuffer

PBS Tween® 20 blot wash buffer is a 20X concentration with 1% Tween® 20.


  • Prevents high background by efficiently removing unbound reagents
  • Convenient liquid concentrate for blotting and ELISA applications


pH (1:20, water) @ 25 °C is 7,50 to 7,70.


Concentrate (500 ml) prepares 10 litres of a 1X buffer.


When diluted down to a 1X concentration, 0,05% Tween® 20 is present.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR