Broxettes of evaporated salt tablets
Lieferant: KAMMERER
1161131
KAMM1161131EA 29.5 EUR
KAMM1161131
Broxettes of evaporated salt tablets
Reinigungsmittel Waschmittel und Oberflächenreiniger
Pressed evaporated salt tablets for the regeneration of ion exchangers.
  • Highest degree of purity
  • Tested and suitable for chlorine chlorinators (according to EN 973 type A)