Replacement Set for Micropipette FX 5/FX 10
Lieferant: KARTELL
0135000
KART0135000EA 68.9 EUR
KART0135000
Replacement Set for Micropipette FX 5/FX 10
Pipetten Mikropipetten
Spare/replacement part set for the micropipette FX 5/FX 10 series pipettors.
Set includes 1 cone, 2 PTFE seals, 2 O-rings, and 1 cone nut.