Sterile Pipette Tips in Rack for Gel Loading
Lieferant: KARTELL
0984600
KART0984600EA 842 EUR
KART0984600
Sterile Pipette Tips in Rack for Gel Loading
Pipettenspitzen
These tips are made of high quality, 100% pure, virgin PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • High clarity, excellent water-repellent and low retention properties
  • Precise centering of the orifice guarantees reliable directionality of the sample when delivered
  • Able to collect a human biological sample and dispense on growth medium, slide tube/containers to carry out analysis of laboratory diagnosis
To be used whenever it is difficult to reach the bottom of test tubes with standard products.