Ice bucket, with lid
Lieferant: KARTELL
1000
KART1000EA 226 EUR
KART1000
Ice bucket, with lid
Eisbehälter
Made of red PUR. Equipped with lid and two side-handles. Can be used with dry or minced ice and cooling mixtures.
  • Thickness of about 15 mm
  • Excellent isolating feature
  • Liquid nitrogen resistant for a few minutes