Industrieschaufel, plastilab®
Lieferant: KARTELL
1066 1111 1110 1112
KART1066EA 287 EUR
KART1066 KART1111 KART1110 KART1112
Industrieschaufel, plastilab®
Schaufeln
These scoops made from HDPE are designed for heavy industrial use.
  • Round shaped design facilitates complete transfer of sampling material
  • Suitable for foodstuff