Enghalsflaschen
Lieferant: KARTELL
15700 15701 15702 15703 15704
KART15700EA 3.4 EUR
KART15700 KART15701 KART15702 KART15703 KART15704
Enghalsflaschen
Flaschen Kunststoffflaschen
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Suitable for foodstuff
The cylindrical bottles are supplied with insert and cap. Thanks to the large neck they are suitable for powders, solid specimen, creamy or semisolid substances, crystals and hygroscopic substances.
Standards: EC Regulation 1935/2004 • EC Regulation 10/2011