Pipette bulb, with adapter
Lieferant: KARTELL
200
KART200EA 23.4 EUR
KART200
Pipette bulb, with adapter
Pipetten Pipettierbälle
Moulded pipette bulb made of synthetic rubber.
  • Three glass ball valves
  • Can be controlled by finger pressure
  • No metal parts to corrode
  • Can be used with all liquids except those that attack rubber
  • Use on bulb or cylindrical pipettes of different capacities
Lieferumfang: Supplied with adapter for pipettes with capacities more than 20 ml.