Bowl
Lieferant: KARTELL
258
KART258EA 200 EUR
KART258
Bowl
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Made of PP, white.
  • With rim
  • Autoclavable at 121 °C for 20 minutes
  • Sturdy structure
Two grips have been designed in the lower part of the rim to help carriage.