Rods for clot detachment and extraction
Lieferant: KARTELL
2652
KART2652EA 54.6 EUR
KART2652
Rods for clot detachment and extraction
Feste Stäbe
High impact PS, white.
Disposable shock-resistant PS rods for detaching and extracting clots.