Pressure bulbs
Lieferant: KARTELL
3910 3911 3912 3913
KART3910EA 49.4 EUR
KART3910 KART3911 KART3912 KART3913
Pressure bulbs
Pipetten Pipettierbälle
Made of rubber, red.
Mono-balls with valve are suitable for pellet burette charging or for vapourisers.
Double balls with net are suitable for automatic burettes and flash chromatography columns.
Tube length: 33,5 cm
Bestellen
TECHNISCHE DATEN
Environmentally Preferable