Disconnectors, Tapered
Lieferant: KARTELL
7053700
KART7053700EA 3700 EUR
KART7053700
Disconnectors, Tapered
Fittings und Verbinder
These quick disconnector is useful for joining hoses of glass, rubber or plastic. Connectors fit tightly together. Replace expensive adapters.
Dimensions are shown as both nominal and exact as elasticity of hoses will influence the choice of size.
Zubehörinformationen: Tested according to reach regulation 1907/2006 Annex XVII.