Bottle-top dispensers, DOSILab
Lieferant: KARTELL
83082 83083 83084 83085 83086 83087
KART83082EA 742 EUR
KART83082 KART83083 KART83084 KART83085 KART83086 KART83087
Bottle-top dispensers, DOSILab
Flaschenaufsatz-Dispenser
These dosilab series are of adjustable volume bottle top dispenser for dosing chemicals with precision and accuracy. The internal components that in contact with liquid solutions are made of PTFE, FEP and borosilicate glass.
  • Low economic value
  • Autoclavable at +121 °C
  • Calibrated easily with other accessory
  • PTFE body with high chemical resistance
  • PTFE piston with silicone O-ring ensuring a high degree of accuracy
They possess excellent compatibility with all reagents (even organic solvents and concentrated acids), except for hydrofluoric acid.