Rods, glass
Lieferant: Kavalier
271070000
KAVA271070000EA 792 EUR
KAVA271070000
Rods, glass
Feste Stäbe
Borosilicate glass 3.3.
  • Appropriately cut to 1500 mm in length (cut ends)
  • Very good chemical and thermal resistance
  • Permissible diameter tolerance of ±0,95 mm