Trichter, gerippt
Lieferant: Kavalier
632413004100
KAVA632413004100EA 50.6 EUR
KAVA632413004100
Trichter, gerippt
Trichter Filtertrichter Filtertrichter
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With ribbed walls.