Pycnometer, according to Gay-Lussac
Lieferant: Kavalier
632431722019
KAVA632431722019EA 1550 EUR
KAVA632431722019
Pycnometer, according to Gay-Lussac
Pyknometer
SIMAX®, borosilicate glass.
  • Autoclavable at 121 °C
Standards: According to DIN 12801 and ISO 3507