Staining jar according to Schiefferdecker
Lieferant: Kavalier
632499890001
KAVA632499890001EA 86.4 EUR
KAVA632499890001
Staining jar according to Schiefferdecker
Dosen Färbeküvetten
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.