Staining jars acc. to Hellendahl
Lieferant: Kavalier
N23253111
KAVAN23253111EA 76.5 EUR
KAVAN23253111
Staining jars acc. to Hellendahl
Dosen Färbeküvetten
Soda-lime glass.
These staining trough hold up to 16 slides (76×26 mm).