Cold traps, type KF NW
Lieferant: KGW
17380-NW16
KGWA17380-NW16EA 1070 EUR
KGWA17380-NW16
Cold traps, type KF NW
Vakuumfallen
Simple cold traps, which are also known as cold finger condensers, are used in conjunction with vacuum pumps and the condensation Produced from humidity or solvents these cold traps can be used for many different tasks. With glass vacuum flanges you are able to connect this cold trap directly to the metal flanges of a high vacuum pump.
Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Reliable, perfect version, easy to clean and handle
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminum
  • Pressure-free cooling sphere inside the Dewar flask
These simple cold finger condensers are used in standard applications in conjunction with vacuum pumps and the condensation is forced out of the humidity or solvents used in the chemical applications. Cold traps can be changed very quickly without any problems and it can be connected directly to the metal flanges of a high vacuum pump with its glass vacuum flanges.