Sample cups for auto analyser
Lieferant: KIMA SAS DI CHIARIN RENZO
18151 18153 18320
KIMA18151EA 0 EUR
KIMA18151 KIMA18153 KIMA18320
Sample cups for auto analyser
Cups made of PS for auto analyser.
  • Good chemical resistance
  • Good temperature resistance