Microtubes
Lieferant: KIMA SAS DI CHIARIN RENZO
18167
KIMA18167EA 0 EUR
KIMA18167
Microtubes
Röhrchen Mikroröhrchen
PP, translucent.
  • Without cap
  • Excellent chemical resistance
  • Excellent temperature resistance
Conical test tube for autoanalyser Olly type.