Wipe rolls, WYPALL* L20 clean and maintenance
Lieferant: KIMBERLY CLARK
72773
KIMB72773EA 69.3 EUR
KIMB72773
Wipe rolls, WYPALL* L20 clean and maintenance
Wischtücher Wisch- und Handtücher, Einweg Wischtücher für allgemeine Anwendungen
WypAll® wiping paper products are designed for the specific environments where they will be used, to deliver on your wiping task, every time.
  • Portable and durable
  • Embossing technology delivers a great balance of performance and value
These rolls are compatible with the quick-loading, versatile and easy-clean Aquarius™ centrefeed dispensing system (KIMB7017).