Micro-hematocrit capillary tubes, KIMBLE®
Lieferant: KIMBLEKONTES
41B2501
KIML41B2501EA 0 EUR
KIML41B2501
Micro-hematocrit capillary tubes, KIMBLE®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
Soda lime glass, 90 expansion.
  • Packed in plastic vials with reclosable snap-lock caps to maintain cleanliness
  • Ends may be sealed by flame or sealing clay
  • Heparinised capillary tubes are designed to prevent blood clotting
For measuring volume percentage of red blood cells.
Standards: Glass conforms to USP Type III requirements.