Glass cleaner, Suma Rapid D6
Lieferant: KING DISPOSABLES
LE511964
KINGLE511964EA 24.8 EUR
KINGLE511964
Glass cleaner, Suma Rapid D6
Reinigungsmittel Waschmittel und Oberflächenreiniger
Suma Rapid D6 is a quick drying glass cleaner for use on windows, mirrors, display cabinets, tiles and other glazed ceramics.
  • Easy spray-on, wipe-off action
  • Quick drying with no residues
  • Leaves glass sparkling with no streaks and smears
Standards: Conforms to SABS 1828 'Cleaning Chemicals for use in the food industry'.