Dialysemembranen, Spectra/Por® 4
Lieferant: KLEINFELD
1216004
KLEI1216004EA 584 EUR
KLEI1216004
Dialysemembranen, Spectra/Por® 4
Schläuche Dialyseschläuche
These membranes, made of regenerated cellulose, contain small amounts of impurities such as heavy metals and sulphides. They are delivered dry, but are protected by a glycerin layer from drying.
  • High resistance to organic solvents
  • Heat resistant up to 60 °C
  • pH range 2 to 12
These membranes, made of regenerated cellulose, contain small amounts of impurities such as heavy metals and sulphides. They are delivered dry, but are protected by a glycerin layer from drying.