Storage Boxes for Blocks
Lieferant: KLINIPATH
1606
KLIN1606EA 172 EUR
KLIN1606
Storage Boxes for Blocks
Boxen Lagerboxen
This storage box is complete with base and lid with writing area.
  • Made of white cardboard
  • Capacity approx. 250 blocks
The box is delivered without divider but with cap.