Syringe for vapour pressure osmometer
Lieferant: KNAUER
A0433
KNAUA0433EA 800 EUR
KNAUA0433
Syringe for vapour pressure osmometer
Spritzen Spritzen für die Chromatographie
This syringe has a glass barrel, stainless steel piston with PTFE seal, including needle with spring stopper.