Isolation Amplifier
Lieferant: KNICK
P27058F1
KNICP27058F1EA 614 EUR
KNICP27058F1
Isolation Amplifier
Füllstandssensoren
Knick P27058 F1 DC isolation amplifier 20 to 253 V.
  • Flexible and extremely precise
  • Broad-range power supply
  • Compact design
  • Quick and easy configuration
  • Pluggable screw terminals
  • 3 port separation
  • Maximum accuracy and specific test report
  • Maximum reliability with five year warranty
The isolation amplifiers provide safe galvanic isolation for any input signals from ±20 mV to ±200 V and from ±0,1 to ±100 mA.