Equipotential bonding electrode
Lieferant: KNICK
ZU0073
KNICZU0073EA 498 EUR
KNICZU0073
Equipotential bonding electrode
Elektroden Elektroden, sonstige
Electrode for monitoring the reference system of the pH sensor or for simultaneous ORP measurements.