Membrane kit for O₂ electrodes
Lieferant: KNICK
ZU0564
KNICZU0564EA 509 EUR
KNICZU0564
Membrane kit for O₂ electrodes
Filter Membranfilterkapseln
This kit includes: membrane body (4×), O-ring set (1×), and 25 ml electrolyte (1×). Suitable for SE 703/706/707/706 Memosens sensors.