Nail brush, hard bristles
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
2-7591
KOEH2-7591EA 12.9 EUR
KOEH2-7591
Nail brush, hard bristles
Bürsten
This nail brush is suitable for general cleaning applications.
  • Autoclavable at 121 °C
  • Cold resistant to –20 °C
  • Hole for hanging
  • Hard bristles