Pipette stand
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
A-1650
KOEHA-1650EA 312 EUR
KOEHA-1650
Pipette stand
Pipettenständer
Universal pipette stand made of rigid PVC. Accommodates up to 9 µl pipettes from various manufacturers.
  • Stable
  • Ergonomically tilted