Injection stoppers
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
K-1240
KOEHK-1240EA 37.3 EUR
KOEHK-1240
Injection stoppers
Stopfen Gummistopfen
These cylindrical shaped stoppers are made of chlorobutyl rubber and are used for injection bottles.
  • Max. temp. 150 °C