Pipette Boxes, Square
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
W-1154 W-1155 W-1153 W-1152 W-1151 W-1150
612-2044EA 112 EUR
612-2044 612-2045 612-2043 612-2042 612-2041 KOEHW-1150
Pipette Boxes, Square
Boxen Pipetten-Sterilisationsbehälter
Anodized aluminium boxes with silicone insert for the protective storage of pipettes.
  • With slip-on cover
  • Autoclavable up to 180 °C