Weighing dishes
Lieferant: Korff
60700
KORF60700EA 59.3 EUR
KORF60700
Weighing dishes
Schiffchen Wägeschiffchen
Dishes, made of aluminium.
  • Lightweight
  • Good temperature resistance
  • Good chemical resistance
Suitable for moisture determination. Thickness: 0,1 mm.