Orcein

Lieferant: SERACARE LIFE SCIENCES

Synonyme: Naturrot 28

1971-01-01
KPLI1971-01-01EA 0 EUR
KPLI1971-01-01
Orcein
Orcein
MDL: MFCD00062310
CAS-Nummer: 1400-62-0
EINECS: 215-750-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR