SureBlue™ TMB 1-Component Microwell Peroxidase Substrate, KPL
Lieferant: SERACARE LIFE SCIENCES
5120-0075
KPLI5120-0075EA
178
EUR
KPLI5120-0075
SureBlue™ TMB 1-Component Microwell Peroxidase Substrate, KPL
Tests
ELISAs
SureBlue™ TMB Microwell substrate develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm and turns yellow when stopped by acidification and read at 450 nm.
Recommended for ELISA. Single component, ready to use format of 3,3',5,5' - Tetramethylbenzidine. No reagent preparation required, very convenient.